siRNA Details
| Gene Symbol: C16orf54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175900.3 | NP_787096.2 | 2 | 1652 | ||
| GenBank mRNA | AK093000.1 | - | 2 | 1639 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C16orf54| Gene Symbol: C16orf54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175900.3 | NP_787096.2 | 2 | 1652 | ||
| GenBank mRNA | AK093000.1 | - | 2 | 1639 | ||