siRNA Details
| Gene Symbol: LOC728095 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109897.1 | 1 | 92 | |||
| GenBank mRNA | AK098705.1 | - | 1 | 55 | ||
| DB028580.1 | - | 1 | 94 | |||
| DB036245.1 | - | 1 | 71 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC728095| Gene Symbol: LOC728095 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109897.1 | 1 | 92 | |||
| GenBank mRNA | AK098705.1 | - | 1 | 55 | ||
| DB028580.1 | - | 1 | 94 | |||
| DB036245.1 | - | 1 | 71 | |||