siRNA Details
| Gene Symbol: LOC101928569 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104669.1 | 3, 4 | 246 | |||
| GenBank mRNA | DB020668.1 | - | 3, 4 | 246 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928569| Gene Symbol: LOC101928569 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104669.1 | 3, 4 | 246 | |||
| GenBank mRNA | DB020668.1 | - | 3, 4 | 246 | ||