siRNA Details
| Gene Symbol: MED14OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289773.1 | NP_001276702.1 | 1 | 323 | ||
| GenBank mRNA | BC039399.1 | - | 1 | 319 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MED14OS| Gene Symbol: MED14OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289773.1 | NP_001276702.1 | 1 | 323 | ||
| GenBank mRNA | BC039399.1 | - | 1 | 319 | ||