siRNA Details
| Gene Symbol: ERVMER34-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242690.1 | NP_001229619.1 | 2 | 311 | ||
| NM_024534.5 | NP_078810.1 | 2 | 311 | |||
| GenBank mRNA | AY189288.1 | - | 2 | 71 | ||
| BP871936.1 | - | 2 | 311 | |||
| DA633167.1 | - | 2 | 311 | |||

ERVMER34-1
Genomic Map