siRNA Details
| Gene Symbol: ERVMER34-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242690.1 | NP_001229619.1 | 2, 3 | 439 | ||
| GenBank mRNA | DA633167.1 | - | 2, 3 | 439 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ERVMER34-1| Gene Symbol: ERVMER34-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242690.1 | NP_001229619.1 | 2, 3 | 439 | ||
| GenBank mRNA | DA633167.1 | - | 2, 3 | 439 | ||