siRNA Details
| Gene Symbol: RSPH10B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099697.1 | NP_001093167.1 | 7 | 867 | ||
| XM_006715766.2 | XP_006715829.1 | 8 | 1004 | |||
| XM_011515491.1 | XP_011513793.1 | 7 | 896 | |||
| XM_011515493.1 | XP_011513795.1 | 7 | 829 | |||
| GenBank mRNA | AK093411.1 | - | 2 | 204 | ||
| AK314983.1 | - | 6 | 987 | |||
| BC044242.1 | - | 3 | 194 | |||
| Gene Symbol: RSPH10B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173565.3 | NP_775836.3 | 7 | 867 | ||
| XM_005249659.3 | XP_005249716.1 | 7 | 1369 | |||
| XM_011515203.1 | XP_011513505.1 | 8 | 949 | |||
| XM_011515204.2 | XP_011513506.1 | 7 | 1137 | |||
| XM_011515207.1 | XP_011513509.1 | 7 | 829 | |||
| XM_011515208.2 | XP_011513510.1 | 7 | 1369 | |||
| XM_011515209.2 | XP_011513511.1 | 7 | 1369 | |||
| XM_011515212.2 | XP_011513514.1 | 7 | 1369 | |||
| XM_017011870.1 | XP_016867359.1 | 7 | 1369 | |||
| GenBank mRNA | BC034495.1 | - | 7 | 870 | 4825868 | |
| BC157864.1 | - | 5 | 617 | 9057083 | ||
| BC157871.1 | - | 5 | 617 | 9057090 | ||

RSPH10B2
Genomic Map