siRNA Details
| Gene Symbol: UBAC2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036531.1 | 1, 2 | 391 | |||
| GenBank mRNA | AK097488.1 | - | 1, 2 | 396 | ||
| DB032212.1 | - | 1, 2 | 392 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

UBAC2-AS1| Gene Symbol: UBAC2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036531.1 | 1, 2 | 391 | |||
| GenBank mRNA | AK097488.1 | - | 1, 2 | 396 | ||
| DB032212.1 | - | 1, 2 | 392 | |||