siRNA Details
| Gene Symbol: UBAC2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036531.1 | 2 | 441 | |||
| NR_036532.1 | 2 | 167 | ||||
| GenBank mRNA | AK097488.1 | - | 2 | 446 | ||
| BC043515.1 | - | 2 | 122 | |||
| DB032212.1 | - | 2 | 442 | |||
| DB446060.1 | - | 2 | 167 | |||

UBAC2-AS1
Genomic Map