siRNA Details
| Gene Symbol: FAM155A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080396.2 | NP_001073865.1 | 2 | 1536 | ||
| XM_011521109.2 | XP_011519411.1 | 2 | 1679 | |||
| GenBank mRNA | AK295445.1 | - | 2 | 1242 | ||
| BC146878.1 | - | 2 | 1137 | 9056806 | ||
| BC157830.1 | - | 2 | 1137 | 9057049 | ||
| BC157855.1 | - | 2 | 1137 | 9057074 | ||
| BC157862.1 | - | 2 | 1137 | 9057081 | ||
| L10374.1 | - | 2 | 1171 | |||

FAM155A
Genomic Map