siRNA Details
| Gene Symbol: CARMN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105059.1 | 4 | 605 | |||
| NR_105060.1 | 3 | 541 | ||||
| GenBank mRNA | AK126481.1 | - | 5 | 1029 | ||
| AL137734.1 | - | 1 | 1366 | |||
| CR933654.1 | - | 2 | 3995 | |||
| HQ848934.1 | - | 4 | 605 | |||
| HQ848935.1 | - | 3 | 541 | |||

CARMN
Genomic Map