siRNA Details
| Gene Symbol: NRBF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282405.1 | NP_001269334.1 | 3 | 1645 | ||
| NM_030759.4 | NP_110386.2 | 4 | 1730 | |||
| XM_006717810.3 | XP_006717873.1 | 5 | 1915 | |||
| GenBank mRNA | AK054957.1 | - | 4 | 1688 | ||
| AK225256.1 | - | 4 | 1637 | |||
| AK225257.1 | - | 4 | 1639 | |||
| AK301656.1 | - | 3 | 1567 | |||
| AL136656.1 | - | 4 | 1685 | |||
| AY267839.1 | - | 5 | 1469 | |||
| AY267840.1 | - | 4 | 1616 | |||
| BC001345.1 | - | 1 | 1314 | |||
| BC011707.1 | - | 4 | 1604 | 3687848 | ||
| DB522359.1 | - | 1 | 275 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001036293, BC019448, AK004200, AK002810.

NRBF2
Genomic Map