siRNA Details
| Gene Symbol: LOC441081 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073404.1 | 1 | 2335 | |||
| GenBank mRNA | BI061609.1 | - | 1 | 288 | ||
| DC422530.1 | - | 1 | 195 | |||
| Gene Symbol: LOC100652833 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011543783.2 | XP_011542085.1 | 1 | 2189 | ||
| Gene Symbol: LOC101929599 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017010105.1 | XP_016865594.1 | 1 | 409 | ||
| Gene Symbol: LOC107986351 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017010104.1 | XP_016865593.1 | 1 | 403 | ||
| Gene Symbol: LOC728488 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK296222.1 | - | 1 | 736 | ||
| AK311627.1 | - | 1 | 196 | |||

LOC441081
Genomic Map