siRNA Details
| Gene Symbol: ZNF727 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159522.1 | NP_001152994.1 | 4 | 644 | ||
| XM_017012225.1 | XP_016867714.1 | 3 | 1185 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF727| Gene Symbol: ZNF727 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159522.1 | NP_001152994.1 | 4 | 644 | ||
| XM_017012225.1 | XP_016867714.1 | 3 | 1185 | |||