siRNA Details
| Gene Symbol: MTUS2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046378.1 | 1, 2 | 256 | |||
| GenBank mRNA | BC027486.1 | - | 1, 2 | 256 | ||
| BI223246.1 | - | 1, 2 | 255 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MTUS2-AS1| Gene Symbol: MTUS2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046378.1 | 1, 2 | 256 | |||
| GenBank mRNA | BC027486.1 | - | 1, 2 | 256 | ||
| BI223246.1 | - | 1, 2 | 255 | |||