siRNA Details
| Gene Symbol: LOC728485 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001294306.1 | NP_001281235.1 | 1 | 107 | ||
| GenBank mRNA | AK092138.1 | - | 1 | 73 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC728485| Gene Symbol: LOC728485 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001294306.1 | NP_001281235.1 | 1 | 107 | ||
| GenBank mRNA | AK092138.1 | - | 1 | 73 | ||