siRNA Details
| Gene Symbol: LOC441081 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073404.1 | 1 | 1444 | |||
| Gene Symbol: LOC646652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | U21556.1 | - | 1 | 381 | ||

LOC441081
Genomic Map