siRNA Details
| Gene Symbol: CRTC3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120372.1 | 2 | 185 | |||
| GenBank mRNA | BG472948.1 | - | 2 | 186 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CRTC3-AS1| Gene Symbol: CRTC3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120372.1 | 2 | 185 | |||
| GenBank mRNA | BG472948.1 | - | 2 | 186 | ||