siRNA Details
| Gene Symbol: LINC01125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038386.1 | 8 | 774 | |||
| GenBank mRNA | AA448462.1 | - | 2 | 142 | ||
| BC039374.1 | - | 8 | 772 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01125| Gene Symbol: LINC01125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038386.1 | 8 | 774 | |||
| GenBank mRNA | AA448462.1 | - | 2 | 142 | ||
| BC039374.1 | - | 8 | 772 | |||