siRNA Details
Gene Symbol: KLHL17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006710600.3 | XP_006710663.1 | 1 | 99 | ||
XM_006710601.3 | XP_006710664.1 | 1 | 99 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KLHL17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006710600.3 | XP_006710663.1 | 1 | 99 | ||
XM_006710601.3 | XP_006710664.1 | 1 | 99 |