siRNA Details
| Gene Symbol: DCDC2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287444.1 | NP_001274373.1 | 5 | 722 | ||
| XM_006711894.3 | XP_006711957.1 | 5 | 716 | |||
| XM_011510393.2 | XP_011508695.1 | 5 | 716 | |||
| XM_017004836.1 | XP_016860325.1 | 5 | 716 | |||
| XM_017004837.1 | XP_016860326.1 | 5 | 716 | |||
| XM_017004838.1 | XP_016860327.1 | 4 | 587 | |||
| GenBank mRNA | DW010024.1 | - | 5 | 417 | ||

DCDC2C
Genomic Map