siRNA Details
| Gene Symbol: PTDSS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290225.1 | NP_001277154.1 | 1 | 309 | ||
| NM_014754.2 | NP_055569.1 | 1 | 309 | |||
| GenBank mRNA | AK293036.1 | - | 1 | 293 | ||
| BC002376.2 | - | 1 | 282 | 2961075 | ||
| BC004192.2 | - | 1 | 259 | 3509418 | ||
| BC004390.1 | - | 1 | 259 | 3634879 | ||
| D14694.1 | - | 1 | 256 | |||
| HY087183.1 | - | 1 | 309 | |||

PTDSS1
Genomic Map