siRNA Details
| Gene Symbol: TMEM109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024092.2 | NP_076997.1 | 1 | 278 | ||
| GenBank mRNA | AK303961.1 | - | 1 | 224 | ||
| AK313913.1 | - | 1 | 60 | |||
| BI463800.1 | - | 1 | 309 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM109| Gene Symbol: TMEM109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024092.2 | NP_076997.1 | 1 | 278 | ||
| GenBank mRNA | AK303961.1 | - | 1 | 224 | ||
| AK313913.1 | - | 1 | 60 | |||
| BI463800.1 | - | 1 | 309 | |||