siRNA Details
| Gene Symbol: MEIKIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303622.1 | NP_001290551.1 | 3 | 407 | ||
| GenBank mRNA | AB987829.1 | - | 3 | 214 | ||
| DB455285.1 | - | 3 | 407 | |||
| DY655061.1 | - | 2 | 92 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MEIKIN| Gene Symbol: MEIKIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303622.1 | NP_001290551.1 | 3 | 407 | ||
| GenBank mRNA | AB987829.1 | - | 3 | 214 | ||
| DB455285.1 | - | 3 | 407 | |||
| DY655061.1 | - | 2 | 92 | |||