siRNA Details
| Gene Symbol: DNHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144666.2 | NP_653267.2 | 31 | 10467 | ||
| GenBank mRNA | AB231767.1 | - | 1 | 761 | ||
| AB231769.1 | - | 4 | 786 | |||
| AB231775.1 | - | 1 | 755 | |||
| AB231777.1 | - | 6 | 716 | |||
| AB290171.1 | - | 18 | 7167 | |||

DNHD1
Genomic Map