siRNA Details
Gene Symbol: LINC01346 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040065.1 | 2 | 192 | |||
GenBank mRNA | AK124708.1 | - | 2 | 187 | ||
DB288420.1 | - | 2 | 239 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01346 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040065.1 | 2 | 192 | |||
GenBank mRNA | AK124708.1 | - | 2 | 187 | ||
DB288420.1 | - | 2 | 239 |