siRNA Details
| Gene Symbol: ZBED3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024398.1 | 3 | 424 | |||
| GenBank mRNA | AK055144.1 | - | 4 | 847 | ||
| AK311179.1 | - | 5 | 606 | |||
| CX786597.1 | - | 3 | 481 | |||
| DA293448.1 | - | 4 | 461 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZBED3-AS1| Gene Symbol: ZBED3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024398.1 | 3 | 424 | |||
| GenBank mRNA | AK055144.1 | - | 4 | 847 | ||
| AK311179.1 | - | 5 | 606 | |||
| CX786597.1 | - | 3 | 481 | |||
| DA293448.1 | - | 4 | 461 | |||