siRNA Details
| Gene Symbol: ANKRD20A8P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | CR936649.1 | - | 2 | 2375 | ||
| Gene Symbol: ANKRD20A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098805.1 | NP_001092275.1 | 15 | 3361 | ||

ANKRD20A8P
Genomic Map