siRNA Details
| Gene Symbol: SLC22A25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017017684.1 | XP_016873173.1 | 2 | 171 | ||
| XM_017017685.1 | XP_016873174.1 | 2 | 171 | |||
| XM_017017687.1 | XP_016873176.1 | 2 | 171 | |||
| XM_017017690.1 | XP_016873179.1 | 2 | 171 | |||
| XM_017017692.1 | XP_016873181.1 | 2 | 171 | |||
| XM_017017695.1 | XP_016873184.1 | 2 | 171 | |||
| XM_017017696.1 | XP_016873185.1 | 2 | 171 | |||

SLC22A25
Genomic Map