siRNA Details
| Gene Symbol: ZNF736 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170905.2 | NP_001164376.1 | 4 | 1008 | ||
| GenBank mRNA | AK301806.1 | - | 4 | 852 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF736| Gene Symbol: ZNF736 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170905.2 | NP_001164376.1 | 4 | 1008 | ||
| GenBank mRNA | AK301806.1 | - | 4 | 852 | ||