siRNA Details
| Gene Symbol: KRTAP1-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030966.1 | NP_112228.1 | 1 | 626 | ||
| GenBank mRNA | AJ406927.1 | - | 1 | 626 | ||
| BC069161.1 | - | 1 | 625 | 7216931 | ||
| BC101150.1 | - | 1 | 620 | 40015405 | ||
| BC101151.2 | - | 1 | 620 | 40015406 | ||
| BC101152.1 | - | 1 | 650 | 40015409 | ||
| BC101153.2 | - | 1 | 620 | 40015410 | ||

KRTAP1-3
Genomic Map