siRNA Details
Gene Symbol: ZNF705D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039615.3 | NP_001034704.2 | 7 | 906 | ||
XM_011543832.2 | XP_011542134.1 | 5 | 688 | |||
GenBank mRNA | BC110823.1 | - | 7 | 906 | 6164596 |
Gene Symbol: ZNF705B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193630.1 | NP_001180559.1 | 7 | 766 | ||
GenBank mRNA | AK292586.1 | - | 7 | 766 |
