siRNA Details
Gene Symbol: UNC5B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038453.1 | 2 | 587 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: UNC5B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038453.1 | 2 | 587 |