siRNA Details
| Gene Symbol: RPEL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143909.1 | NP_001137381.1 | 1 | 1304 | ||
| GenBank mRNA | BC157875.1 | - | 1 | 1305 | 9057093 | |
| BC171885.1 | - | 1 | 1306 | 9054539 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPEL1| Gene Symbol: RPEL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143909.1 | NP_001137381.1 | 1 | 1304 | ||
| GenBank mRNA | BC157875.1 | - | 1 | 1305 | 9057093 | |
| BC171885.1 | - | 1 | 1306 | 9054539 | ||