siRNA Details
| Gene Symbol: NCK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004720.2 | NP_001004720.1 | 4 | 1780 | ||
| NM_001004722.3 | NP_001004722.1 | 4 | 1168 | |||
| NM_003581.4 | NP_003572.2 | 5 | 1890 | |||
| XM_006712797.3 | XP_006712860.1 | 5 | 2546 | |||
| XM_011511991.2 | XP_011510293.1 | 5 | 1984 | |||
| XM_011511992.2 | XP_011510294.2 | 3 | 1112 | |||
| XM_017005103.1 | XP_016860592.1 | 6 | 2148 | |||
| XM_017005104.1 | XP_016860593.1 | 5 | 2160 | |||
| XM_017005105.1 | XP_016860594.1 | 5 | 1897 | |||
| GenBank mRNA | BC000103.2 | - | 4 | 1664 | 3505427 | |
| BC007195.1 | - | 4 | 1671 | 3608149 | ||

NCK2
Genomic Map