siRNA Details
Gene Symbol: FAAHP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045483.1 | 7, 8 | 625 | |||
GenBank mRNA | BI756183.1 | - | 4, 5 | 316 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAAHP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045483.1 | 7, 8 | 625 | |||
GenBank mRNA | BI756183.1 | - | 4, 5 | 316 |