siRNA Details
| Gene Symbol: LINC01630 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040074.1 | 1 | 196 | |||
| NR_040075.1 | 1 | 196 | ||||
| GenBank mRNA | AK094936.1 | - | 1 | 196 | ||
| AK294844.1 | - | 1 | 196 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01630| Gene Symbol: LINC01630 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040074.1 | 1 | 196 | |||
| NR_040075.1 | 1 | 196 | ||||
| GenBank mRNA | AK094936.1 | - | 1 | 196 | ||
| AK294844.1 | - | 1 | 196 | |||