siRNA Details
| Gene Symbol: OIP5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026757.1 | 2 | 187 | |||
| GenBank mRNA | AK172729.1 | - | 2 | 182 | ||
| AY007106.1 | - | 2 | 169 | |||
| BC037835.1 | - | 2 | 218 | |||
| BX647764.1 | - | 2 | 186 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OIP5-AS1| Gene Symbol: OIP5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026757.1 | 2 | 187 | |||
| GenBank mRNA | AK172729.1 | - | 2 | 182 | ||
| AY007106.1 | - | 2 | 169 | |||
| BC037835.1 | - | 2 | 218 | |||
| BX647764.1 | - | 2 | 186 | |||