siRNA Details
| Gene Symbol: FAM198A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129908.2 | NP_001123380.2 | 2 | 1087 | ||
| GenBank mRNA | AK311059.1 | - | 2 | 760 | ||
| AL117530.1 | - | 1 | 330 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM198A| Gene Symbol: FAM198A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129908.2 | NP_001123380.2 | 2 | 1087 | ||
| GenBank mRNA | AK311059.1 | - | 2 | 760 | ||
| AL117530.1 | - | 1 | 330 | |||