siRNA Details
| Gene Symbol: STXBP5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC035182.2 | - | 11 | 1202 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

STXBP5-AS1| Gene Symbol: STXBP5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC035182.2 | - | 11 | 1202 | ||