siRNA Details
| Gene Symbol: DNM1P46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003260.1 | 2 | 231 | |||
| GenBank mRNA | AK127834.1 | - | 2 | 231 | ||
| BC101080.2 | - | 2 | 119 | |||
| BC101081.2 | - | 2 | 119 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DNM1P46| Gene Symbol: DNM1P46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003260.1 | 2 | 231 | |||
| GenBank mRNA | AK127834.1 | - | 2 | 231 | ||
| BC101080.2 | - | 2 | 119 | |||
| BC101081.2 | - | 2 | 119 | |||