siRNA Details
| Gene Symbol: PRSS46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205271.1 | NP_001192200.1 | 3 | 392 | ||
| GenBank mRNA | DB058077.1 | - | 3 | 305 | ||
| DB099248.1 | - | 3 | 388 | |||
| DY654973.1 | - | 1 | 74 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRSS46| Gene Symbol: PRSS46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205271.1 | NP_001192200.1 | 3 | 392 | ||
| GenBank mRNA | DB058077.1 | - | 3 | 305 | ||
| DB099248.1 | - | 3 | 388 | |||
| DY654973.1 | - | 1 | 74 | |||