siRNA Details
| Gene Symbol: OC90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080399.2 | NP_001073868.2 | 11 | 853 | ||
| GenBank mRNA | AK297911.1 | - | 12 | 853 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OC90| Gene Symbol: OC90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080399.2 | NP_001073868.2 | 11 | 853 | ||
| GenBank mRNA | AK297911.1 | - | 12 | 853 | ||