siRNA Details
| Gene Symbol: PLIN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080400.1 | NP_001073869.1 | 3 | 743 | ||
| XM_006722866.2 | XP_006722929.1 | 5 | 2704 | |||
| XM_006722868.3 | XP_006722931.1 | 5 | 2700 | |||
| XM_011528233.2 | XP_011526535.1 | 6 | 2886 | |||
| XM_017027192.1 | XP_016882681.1 | 6 | 2889 | |||
| XM_017027193.1 | XP_016882682.1 | 6 | 2836 | |||
| XM_017027194.1 | XP_016882683.1 | 6 | 2871 | |||
| GenBank mRNA | AB067468.1 | - | 519 | |||

PLIN4
Genomic Map