siRNA Details
| Gene Symbol: LMO7DN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001257995.1 | NP_001244924.1 | 2 | 245 | ||
| XM_011535208.2 | XP_011533510.1 | 3 | 2072 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LMO7DN| Gene Symbol: LMO7DN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001257995.1 | NP_001244924.1 | 2 | 245 | ||
| XM_011535208.2 | XP_011533510.1 | 3 | 2072 | |||