siRNA Details
| Gene Symbol: GP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004488.2 | NP_004479.1 | 2 | 504 | ||
| GenBank mRNA | L11238.1 | - | 1 | 545 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GP5| Gene Symbol: GP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004488.2 | NP_004479.1 | 2 | 504 | ||
| GenBank mRNA | L11238.1 | - | 1 | 545 | ||