siRNA Details
| Gene Symbol: CCDC61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267723.1 | NP_001254652.1 | 5 | 511 | ||
| XM_005259191.3 | XP_005259248.1 | 5 | 2616 | |||
| XM_005259192.4 | XP_005259249.1 | 5 | 1257 | |||
| XM_011527257.2 | XP_011525559.1 | 5 | 540 | |||
| GenBank mRNA | BI819372.1 | - | 4 | 411 | ||
| DR007040.1 | - | 5 | 531 | |||
| GQ375050.1 | - | 5 | 484 | |||

CCDC61
Genomic Map