siRNA Details
| Gene Symbol: CCDC61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267723.1 | NP_001254652.1 | 3 | 219 | ||
| XM_005259191.3 | XP_005259248.1 | 3 | 2324 | |||
| XM_005259192.4 | XP_005259249.1 | 3 | 965 | |||
| XM_011527257.2 | XP_011525559.1 | 3 | 248 | |||
| GenBank mRNA | DR007040.1 | - | 3 | 239 | ||
| GQ375050.1 | - | 3 | 192 | |||

CCDC61
Genomic Map