siRNA Details
| Gene Symbol: GPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001257199.1 | NP_001244128.1 | 2 | 408 | ||
| NM_005276.3 | NP_005267.2 | 2 | 408 | |||
| GenBank mRNA | AK130162.1 | - | 2 | 219 | ||
| AK295911.1 | - | 2 | 205 | |||
| AK312516.1 | - | 2 | 190 | |||
| BC032234.1 | - | 2 | 209 | 5229925 | ||
| DC305085.1 | - | 2 | 381 | |||
| DC310521.1 | - | 2 | 411 | |||
| L34041.1 | - | 2 | 205 | |||

GPD1
Genomic Map