siRNA Details
| Gene Symbol: TMEM242 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018452.5 | NP_060922.2 | 3 | 1078 | ||
| GenBank mRNA | AF161405.1 | - | 3 | 415 | ||
| AF217510.1 | - | 3 | 420 | |||
| AK000891.1 | - | 3 | 326 | |||
| BC029130.1 | - | 3 | 346 | 5182086 | ||
| BC146893.1 | - | 3 | 388 | 9056821 | ||
| BC146895.1 | - | 3 | 388 | 9056823 | ||

TMEM242
Genomic Map